Order Entry
Italy
ContactUsLinkComponent
DNA (Deoxyribonucleic acid) (from salmon testes)
DNA (Deoxyribonucleic acid) (from salmon testes)
Catalog # ROCKMB-103-1000
CAS Number:  
DNA (Deoxyribonucleic acid) (from salmon testes)
Catalog # ROCKMB-103-1000
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
CAS Number:9007-49-2
 :100403-24-5
MDL Number:MFCD00130922