Biohazard Bags
Catalog # GREI649203_350
Supplier: Greiner Bio-One
Specifications
- Packed:300 pcs/pack
- Capacity:130 L
- Description:PP disposal bag with biohazard imprint, 69 g
- L×W:700×1100 mm
- Material:PP
- Seq:02
- Pk:350
Specifications
About this item
PP disposal bag, with imprint 'Biohazard', for single use
- Translucent bag with red printing
- Suitable for steam autoclaves (max. 134 °C, 2 bar/30 min)
- Temperature range: 0 to 140 °C