Latex gloves, TouchNTuff® 69-318
Supplier: Ansell
Certificates
About this item
These ambidextrous gloves are resistant to splashes of acids, bases, alcohols and dilute aqueous solutions of most chemicals.
- Flexible and comfortable to wear
- Cooler than vinyl, with superior finger flexibility and grip
- Available in both powdered and powder-free versions
- Powdered gloves have a smooth surface, powder-free gloves have a textured finish
Ideal for use in laboratory analysis, electronic manufacturing and in pharmaceutical industries.
EN 374, Cat. III; AQL 1,5 Food contact approved
Certifications: EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS; Cat. III; AQL 1,5
Food contact approved
Food contact approved
Specifications
- Catalog No:
- 112-4265
- 112-4264
- 112-4262
- 112-4266
- 112-4263
- Model:
- 69-318
- 69-318
- 69-318
- 69-318
- 69-318
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Textured
- Textured
- Textured
- Textured
- Textured
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- EN norm:
- EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS
- EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS
- EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS
- EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS
- EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS
- Food contact approved:
- +
- +
- +
- +
- +