Waste disposal bags, Swirl®
Elkay's water-soluble and eco-friendly bags are ideal for minimizing employee exposure to contaminated laundry.
- Protects personnel from contamination and dissolves completely at a water temperature of 140 ° F or above
: The binding tape is attached directly to the bag for a simple closure. Wet or damp laundry can safely be stored in the bag for several hours. Neither the laundry itself, nor the equipment nor the drainage system will be damaged if used correctly by the bags.
Frequently Bought Together