Polyethylene glycol 400, liquid, J.T.Baker®
Catalog # U216-07
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle, amber, narrow mouth in single shipper
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:Passes Test
- Appearance:Passes Test
- Identification (by IR):Passes Test
Specification Test Results
About this item
:
:400 g/mol
Boiling Pt:∼250 °C (1013 hPa)
Melting Pt:4…8 °C
Density:1.13 g/cm³ (20 °C)
:Management of Change (MOC) category = N
Flash Pt:245 °C
Storage Temperature:Ambient
EINECS:500-038-2
CAS Number:25322-68-3
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD00081839