Single channel micro dispensers, fixed volume
: DRUMMOND
Highly accurate micro-dispensers with borosilicate glass bores and stainless steel plunger.
- Overall volumetric accuracy of ±1% (at full volume)
- PTFE tips on all plungers 25 µl or larger
- Precision molded plastic bores on 1000 μl model
Frequently Bought Together