
Barium carbonate 99.0-101.0% (by acidimetry), powder, BAKER ANALYZED® ACS, J.T.Baker®
: Avantor
:513-77-9
:
:BaCO₃
:MFCD00003448
:197.34 g/mol
:13,00972
:1350 °C
:4.30 g/cm³
:Ambient
:208-167-3
:1564
:6.1,III
:GHS06
- Meets ACS Reagent Chemical Requirements:99.0 - 101.0%
- For Laboratory,Research,or Manufacturing Use:≤0.015%
- Assay (BaCO₃) (by acidimetry):99.0 - 101.0 %
- Insoluble in Dilute HCl:≤ 0.015 %
- Chloride (Cl):≤ 0.002 %
- Water-Soluble Titrable Base, meq/g:≤ 0.0020
- Oxidizing Substances (as NO₃):≤ 0.005 %
- Sulfide (S):≤ 0.001 %
- Calcium (Ca):≤ 0.05 %
- Heavy Metals (as Pb):≤ 0.001 %
- Iron (Fe):≤ 0.002 %
- Potassium (K):≤ 0.005 %
- Sodium (Na):≤ 0.010 %
- Strontium (Sr):≤ 0.7 %
- Average Particle Diameter, µm (APD):≤ 5
Frequently Bought Together