Entellan® New rapid mounting medium for microscopy
Catalog # 1.07961.0000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Description:Entellan® New rapid mounting medium
Specifications
About this item
Entellan® New is a new rapid mounting medium for cover slippers and is specially suited for commercial automated mounting instruments that operate with cover slippers.
- Refractive index: 1,49 to 1,50
- Storage temperature: 15 to 25 °C
- Viscosity between 500 and 600 mPas
The narrow viscosity range improves and accelerates the application, as neither the dropping rate nor the volume need to be reset after the first run with a new bottle of Entellan® New.
Certifications: Entellan® New is an IVD registered product and CE certified, thus can be used for clinical diagnostic purposes.