Plasmid purification kit, Axygen® AxyPrep™ Mag
Supplier: Corning
Certificates
About this item
AxyPrep™ Mag Plasmid Purification Kit utilises a magnetic bead technology for high throughput purification of plasmid DNA from E.coli cells. The AxyPrep™ Mag kit can also be used with fosmid and BAC vector-based constructs ranging from 5 to 150 Kb. The system consists of steps of pelleting cells, resuspension, lysis, neutralisation, DNA binding, ethanol washing and plasmid elution. The protocol is automation-compatible.
Specifications
- Method / Format:Magnetic beads
- Prep size:Scalable
- Refine target molecule:Plasmid
- Sample type:Bacteria
- Target molecule:Plasmid
- Description:AxyPrep™ Mag Plasmid Purification Kit
Specifications
Frequently Bought Together