Rotary valves, high pressure
: OMNIFIT
The high pressure manual rotary valves are made from PTFE with a PCTFE rotor with a bore size of 0,8 mm making them ideal for pressure applications up to 500 psi. They are available in four and five port versions. All ports are 1/4"-28 UNF flat-bottom and will accept any 1/4"-28 UNF male fitting. These valves are mountable using the pre-drilled M3 mounting holes.
- Pressure rated up to 500 psi
- Accept 1/4"-28 UNF fittings
- M3 mounting holes
Frequently Bought Together