Petri dish holder
Coated petri dish holder for six 90 mm dishes
Catalog # 391-1150
Supplier: SciLabware
Jump to:
Product Details & Documents
Polyamide epoxy coated steel. Holds up to six 90 mm petri dishes.
- Better heat and chemical resistance than other coatings
- Ideal for small batch incubation
Specifications
- Description:Coated petri dish holder for six 90 mm dishes
- Seq:01