Order Entry
Italy
ContactUsLinkComponent
Dilatometers, small quartz
Dilatometers, small quartz
Catalog # PERC03190461
Supplier:  PerkinElmer
Dilatometers, small quartz
Catalog # PERC03190461
Supplier:  PerkinElmer
Specifications
  • Description:
    Dilatometer kit with small quartz
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
The kit permits the determination of volumetric changes of materials upon melting or upon cooling from a melt. A typical application would be the determination of shrinkage from a mold.
  • Kit includes quartz dilatometer cell, aluminum oxide powder (28 g), quartz sample tube and instruction sheet
  • 0,1 and 1,5 ml capacity can be used with the 15 and 28 mm diameter furnace repectively