Examination gloves, latex, Gentle Skin® Sensitive
These ambidextrous gloves are designed for use in areas with high contamination risks. Ideal for people working in hospitals, care facilities and laboratories.
- Non sterile, powder-free, micro-textured finish
- High elasticity, flexibility and tactility
- ProSlide effect for easy and quick donning
- Beaded cuffs
- Low in proteins and accelerators
Colour: White
EN 455; AQL 1,5
: EN 455; AQL 1,5
- Catalog No:
- ROES1221T-XL
- ROES1221T-M
- ROES1221T-L
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Micro-textured
- Micro-textured
- Micro-textured
- Cuff style:
- Beaded
- Beaded
- Beaded
- EN norm:
- EN 455
- EN 455
- EN 455
Frequently Bought Together