Cleanroom gloves, latex/nitrile/neoprene, AdvanTech 514
Supplier: MAPA
Certificates
About this item
Anatomically designed liquid-proof gloves are made of natural latex blended with neoprene and nitrile. They feature a chlorinated internal surface and an anti-slip finish. The material does not contain any powder, silicone or paraffin.
- Non sterile, powder-free, textured finish
- Unique triple polymer blend: superior protection against chemicals commonly used in controlled environments
- Manufacturing process guarantees a very low level of particles, non-volatile residue and extractables
- Wide cuff for double-gloving, extended length to protect the forearm
- Safe wet grip thanks to embossed finish
Suitable for use in Class 10, ISO 4 or M2.5 cleanrooms.
EN 374, EN 388 (1110); AQL 1.5; Cat. III
Certifications: EN ISO 374-1 Type B (KST), EN ISO 374-5, EN 388:2016 (1110X); AQL 1.5; Cat. III.
Frequently Bought Together