Microinjectors, oocyte pipette
Catalog # DRUM3-000-510-X
Supplier: DRUMMOND
Specifications
- Description:Oocyte microinjection pipette
- Pk:1
Specifications
About this item
Oocyte microinjection pipette with non rotating plunger eliminates tip wobble. Injection volume of 30 nanolitre or more.
- Precise deposition
- Reproducable accuracy
- Drives oil interface with minimal or no overshoot
Delivery: Supplied with 100× 8" (2 cm) glass capillaries.