Order Entry
Italy
ContactUsLinkComponent
4-(Dimethylamino)pyridine 14,5 g/l in THF, VOLUSOL®
Flammable
Health
Irritant
4-(Dimethylamino)pyridine 14,5 g/l in THF, VOLUSOL®
Catalog # VWRS83927.410
Supplier:  VWR Collection
CAS Number:  
Flammable
Health
Irritant
4-(Dimethylamino)pyridine 14,5 g/l in THF, VOLUSOL®
Catalog # VWRS83927.410
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pk:
    10 L
  • Seq:
    5214

Specifications

About this item

 :
 :122.17 g/mol
UN:2056
ADR:3,II
 :GHS02
 :GHS08
 :GHS07
CAS Number:1122-58-3
Formula:C₇H₁₀N₂
MDL Number:MFCD00006418