Chemical resistant overalls, Tychem® 4000 S, model CHZ5 / CHZ6
: DuPont
These garments are made from a lightweight, supple and durable fabric (<700 g per garment) consisting of a chemical barrier film laminated to a heavy duty Tyvek® substrate. They achieve a barrier to permeation by a broad range of inorganic and organic chemicals.
- Hood, double zips and double flaps for greater tightness, outer flap with adhesive tape and integrated chin flap
- Elasticated face, wrists, waist and ankles for an optimal fit, thumb loops to prevent sleeves from riding up
- Stitched and over-taped seams with barrier tape for protection and strength
- Double cuffs for good glove compatibility
- Anti-static treatment (EN 1149-5) on inside
- Available with or without attached socks and boot flaps
Tychem® 4000 S is ideal for chemical handling, environmental clean-up operations and emergency response. It is suitable for use in a variety of industries, including oil and gas, chemical engineering, and for use by hazardous material response teams and other emergency services.
Weight: 124 g/m²
Colour: White
: (EU) 2016/425, Type 3B/4B/5B/6B, EN 1149-5, EN 1073-2, EN 14126, Cat. III
: 20 per box, individually packed.
- Catalog No:
- 113-7970
- 113-7971
- 113-7969
- 113-7966
- 113-7967
- 113-7976
- 113-7968
- 113-7977
- 113-7975
- 113-7972
- 113-7973
- 113-7974
- Model:
- 4000 S
- 4000 S
- 4000 S
- 4000 S
- 4000 S
- 4000 S
- 4000 S
- 4000 S
- 4000 S
- 4000 S
- 4000 S
- 4000 S
- Anti-static:
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- Garment application:
- Chemical protection, Medical, Laboratory, Clinical, Industrial
- Chemical protection, Medical, Laboratory, Clinical, Industrial
- Chemical protection, Medical, Laboratory, Clinical, Industrial
- Chemical protection, Medical, Laboratory, Clinical, Industrial
- Chemical protection, Medical, Laboratory, Clinical, Industrial
- Chemical protection, Medical, Laboratory, Clinical, Industrial
- Chemical protection, Medical, Laboratory, Clinical, Industrial
- Chemical protection, Medical, Laboratory, Clinical, Industrial
- Chemical protection, Medical, Laboratory, Clinical, Industrial
- Chemical protection, Medical, Laboratory, Clinical, Industrial
- Chemical protection, Medical, Laboratory, Clinical, Industrial
- Chemical protection, Medical, Laboratory, Clinical, Industrial
Frequently Bought Together