Order Entry
Italy
ContactUsLinkComponent
Suspended solids colorimeter, MD100
Suspended solids colorimeter, MD100
Catalog # LOVI276150
Supplier:  Lovibond
CAS Number:  
undefined
Suspended solids colorimeter, MD100
Catalog # LOVI276150
Supplier:  Lovibond
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    MD100 for suspended solids
  • Model:
    MD100
  • Wavelength accuracy:
    ±1 nm
  • Automatic switch-off:
    after 10 minutes
  • Display:
    LCD backlit
  • Optics:
    LED, interference filters (IF) 660 nm, Δλ=5 nm
  • Power supply:
    4×AAA
  • Weight:
    260 g
  • W×D×H:
    155×75×35 mm
  • Range:
    0 - 750 mg/l TSS

Specifications

About this item

The MD 100 photometer is a portable handheld instrument with a simple button layout, robust waterproof design and large backlit display that are ideal for testing in the field or laboratory conditions.

  • Accurate (3% FS), reproducible results using high quality interference filter with long-life LED
  • Internal ring memory for 16 data sets
  • Real time clock and date
  • OTZ (One Time Zero setting) on versions except those denoted*
  • Optional infrared interface for transfer of test data to PC or printer
Delivery: Instrument supplied with carry case, 4× AAA batteries, 3× round glass vials with lids, relevant reagents, and instruction manual.