Order Entry
Italy
ContactUsLinkComponent
Industrial balances, Defender™ 3000 Hybrid
Industrial balances, Defender™ 3000 Hybrid
Catalog # OHAU80252917UK
Supplier:  OHAUS
undefined
Industrial balances, Defender™ 3000 Hybrid
Catalog # OHAU80252917UK
Supplier:  OHAUS

Specifications

  • Model:
    D32XW30BR-UK
  • Pan/Platform Size:
    355×305 mm
  • Pk:
    1
  • Readability:
    5 g
  • Weighing cap.:
    30000 g

Specifications

About this item

A range of modular scales for tough industrial environments. Easy to use scales that are ideal for general weighing and simple counting applications in production, packaging, and shipping and receiving areas.

  • IP 66 stainless steel indicator
  • Sturdy painted carbon steel tubular frame with high quality 304 stainless steel top plate
  • Internal rechargeable battery