Order Entry
Italy
Orders LinkContactUsLinkComponent
Screw thread vials
Screw thread vials
Supplier:  DELTALAB
Certificates
Screw thread vials
5 Options Available Below
Product Details & Documents
These flat bottom screw thread vials are made of ultra clear borosilicate glass.
  • Fully autoclavable
 : Caps are supplied separately.
 : Supplied in trays individually shrink-wrapped.
Product Family Options
Product Information
  • Capacity
    Base
    Ø×H
    Material
    Colour
    Cap included
    Cap type
    Cap Size
    Availability
    Price
  • 5,00 ml
    Flat bottom
    18×50 mm
    Glass
    Clear
    No
    Screw
    15-425
    Log in for availability
    Log in for availability
    1 * 250 pezzi
    1 * 250 pezzi - 328,00 €
    328,00 €
    1 * 250 pezzi
       328,00 €
  • 10,00 ml
    Flat bottom
    22×50 mm
    Glass
    Clear
    No
    Screw
    18-400
    Log in for availability
    Log in for availability
    1 * 100 pezzi
    1 * 100 pezzi - 133,00 €
    133,00 €
    1 * 100 pezzi
       133,00 €
  • 20,00 ml
    Flat bottom
    28×65 mm
    Glass
    Clear
    Yes
    Screw, with stopper
    22-400
    Log in for availability
    Log in for availability
    1 * 150 pezzi
    1 * 150 pezzi
  • 20,00 ml
    Flat bottom
    28×65 mm
    Glass
    Clear
    No
    Screw
    22-400
    Log in for availability
    Log in for availability
    1 * 152 pezzi
    1 * 152 pezzi - 325,00 €
    325,00 €
    1 * 152 pezzi
       325,00 €
  • 24,00 ml
    Flat bottom
    28×75 mm
    Glass
    Clear
    No
    Screw
    22-400
    Log in for availability
    Log in for availability
    1 * 100 pezzi
    1 * 100 pezzi - 250,00 €
    250,00 €
    1 * 100 pezzi
       250,00 €
Caps
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Black screw cap with internal silicone joint, 15-425 screw
    Log in for availability
    Log in for availability
    1 * 250 pezzi
    1 * 250 pezzi - 519,00 €
    519,00 €
    1 * 250 pezzi
       519,00 €
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Black screw cap with internal silicone joint, 18-400 screw
    Log in for availability
    Log in for availability
    1 * 250 pezzi
    1 * 250 pezzi - 638,00 €
    638,00 €
    1 * 250 pezzi
       638,00 €
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Black screw cap with internal silicone joint, 22-400 screw
    Log in for availability
    Log in for availability
    1 * 300 pezzi
    1 * 300 pezzi - 967,00 €
    967,00 €
    1 * 300 pezzi
       967,00 €