Order Entry
Italy
ContactUsLinkComponent
Labmobile trolleys, 3 tier
Labmobile trolleys, 3 tier
  GUESS9621
 :  GUEST
Labmobile trolleys, 3 tier
  GUESS9621
 :  GUEST

 

  • Colour:
    White
  • Description:
    Labmobiles trolley, 3 tier, ABS, with 2 drawers (90 mm)
  • Seq:
    03
  • W×D×H:
    430×410×730 mm
  • Pk:
    1
  • Shelves:
    2

 

 

Labmobile three tier trolleys provides a worktop at a standard sitting working height.

  • Provides large storage capacity
  • Easy glide manoeuvrability
  • Easy to clean using wipes or spray
  • Suitable for anaesthetists, optometrists, practice nurses and chiropodists

It is also a practical and versatile under bench unit for laboratories and is helpful as an addition to the doctors office desk.