
9-Bromo-1-nonanol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:55362-80-6
:
Formula:C₉H₁₉BrO
MDL Number:MFCD00063348
:223.15 g/mol
Boiling Pt:125…126 °C (2 mmHg)
Melting Pt:33…35 °C
Flash Pt:109 °C (228 °F)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together