
(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine ≥98.0% (by GC, titration analysis)
: TCI
:23357-52-0
:
:C₁₀H₁₃N
:MFCD00671630
:147.22 g/mol
:77°C
:GHS07
- Appearance:Colorless to Red to Green clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:+26.0 to +30.0 deg (C=2, MeOH)
Frequently Bought Together