


S-Methyl thioacetate ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1534-08-3
:
Formula:C₃H₆OS
MDL Number:MFCD00014989
:90.15 g/mol
Boiling Pt:98°C
Flash Pt:12°C
EINECS:216-252-1
UN:3272
ADR:3,II
:GHS02
:GHS05
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together