
3-Fluoro-2-formylpyridine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:31224-43-8
:
Formula:C₆H₄FNO
MDL Number:MFCD07781234
:125.1 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together