
3-Chloro-4-formylpyridine ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:72990-37-5
:
Formula:C₆H₄ClNO
MDL Number:MFCD06200712
:141.56 g/mol
Boiling Pt:93°C
:GHS07
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:58.0 to 62.0 deg-C
Specification Test Results
Frequently Bought Together