
1-Butyl-3-methylimidazoliumbis(trifluoromethylsulphonyl)imide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:174899-83-3
:
Formula:C₁₀H₁₅F₆N₃O₄S₂
MDL Number:MFCD05664714
Melting Pt:–2°C
:419.37 g/mol
:GHS07
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (HPLC):min. 98.0 area%
- Purity (Ion exchange titration):min. 98.0 %
Specification Test Results
Frequently Bought Together