


4-Bromophenol ≥98.0% (by GC, titration analysis) for biochemical research
Supplier: TCI
Certificates
About this item
CAS Number:106-41-2
:
Formula:C₆H₅BrO
MDL Number:MFCD00002313
:173.01 g/mol
Merck Index:14,01428
Boiling Pt:238°C
Melting Pt:63 °C
Density:~ 1,84 g/cm³ (20 °C)
Flash Pt:≥ 110 °C
Storage Temperature:Ambient
EINECS:203-394-4
UN:3077
ADR:9,III
:GHS07
:GHS08
:GHS09
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:63.0 to 67.0 deg-C
- Chemiluminescence test (Luminol/POD/H2O2):to pass test
Specification Test Results
Frequently Bought Together