
3,3,5-Trimethylcyclohexanol ≥80.0% (by GC) (contains ca. 20% trans- isomer)
: TCI
:116-02-9
:
:C₉H₁₈O
:MFCD00019378
:142.24 g/mol
:86°C
:30…32 °C
:0,9006 g/cm³ (16 °C)
:Refrigerator
:204-122-7
:GHS05
- Appearance:White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
- Purity (GC):min. 80.0 %
- Melting point:32.0 to 36.0 deg-C
Frequently Bought Together