

5-Bromocytosine 98%
Catalog # H61952.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
About this item
:
:190 g/mol
Melting Pt:240…243 °C
EINECS:218-806-8
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:2240-25-7
Formula:C₄H₄BrN₃O
MDL Number:MFCD00056025