

2,4-Dibromothiazole ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4175-77-3
:
Formula:C₃HBr₂NS
MDL Number:MFCD01318994
:242.92 g/mol
Melting Pt:80…84 °C
:GHS05
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together