
3,5-Dimethoxybenzylamine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:34967-24-3
:
Formula:C₉H₁₃NO₂
MDL Number:MFCD00052813
:167.21
Boiling Pt:94…96 °C
Melting Pt:35…37 °C
Density:1.106
Flash Pt:113 °C (235 °F)
Storage Temperature:Ambient
UN:3259
ADR:8,III
:GHS05
Related Information
Frequently Bought Together