
1,5-Difenilcarbazone 40,00% (by HPLC) (contains Diphenylcarbazide)
TCIAD0881-25G
: TCI
:
- Tipo di conf.:Bottiglia di vetro
- Conf:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 40.0 area%
:
:240.26 g/mol
:14,03322
:440 °C (1013 hPa)
:157°C
:Ambiente
:208-698-0
:GHS07
:538-62-5
:C₁₃H₁₂N₄O
:MFCD00003024