

Methylene blue ≥98.0% (by titrimetric analysis) ion association reagent for spectrophotometric analysis
Catalog # TCIAA5105-1G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Green to Dark green powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 % (calcd.on dried substance)
- Lambda max.:661.0 to 665.0 nm (EtOH (50%))
- Absorbance:min. 0.540 (2mg/L, EtOH (50%), 661.0 to 665.0 nm)
- Drying loss:max. 5.0 %
Specification Test Results
About this item
:
:319.86 g/mol
Merck Index:14,06060
Melting Pt:180 °C
Density:1,757 g/cm³ (22 °C)
EINECS:200-515-2
:GHS07
:GHS08
Storage Temperature:Ambient
CAS Number:61-73-4
:MFCD00012111
Formula:C₁₆H₁₈ClN₃S