Order Entry
Italy
ContactUsLinkComponent
Tantalum, gauze woven from wire 100 mesh woven from 0.08mm (0.003in)
Tantalum, gauze woven from wire 100 mesh woven from 0.08mm (0.003in)
Catalog # 46695.FL
CAS Number:  
Tantalum, gauze woven from wire 100 mesh woven from 0.08mm (0.003in)
Catalog # 46695.FL
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    1706
  • Pk Info:
    75×75 mm

Specifications

About this item

 :
 :180.95 g/mol
Merck Index:12,09223
Boiling Pt:5427 °C (1013 hPa)
Melting Pt:2996 °C
Density:16.69 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-13505
CAS Number:7440-25-7
Formula:Ta
MDL Number:MFCD00011252