
D-(-)-Leucinol ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:53448-09-2
:
Formula:C₆H₁₅NO
MDL Number:MFCD00004734
:117.19 g/mol
Boiling Pt:194°C
Density:0,897 g/cm³ (20 °C)
Flash Pt:90°C
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 97.0 %
- Optical purity (GC):min. 99.0 ee%
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
Related Information
Frequently Bought Together