
4-Phenyl-1,3-dioxane ≥98.0% (by GC)
TCIAP0166-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
:
:251°C
:212-244-7
:GHS07
:772-00-9
:C₁₀H₁₂O₂
:MFCD00006568