Order Entry
Italy
ContactUsLinkComponent
4-Methyl-1,3-dioxane ≥99.0% (by GC)
Flammable
Irritant
4-Methyl-1,3-dioxane ≥99.0% (by GC)
Catalog # TCIAM0209-25ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
4-Methyl-1,3-dioxane ≥99.0% (by GC)
Catalog # TCIAM0209-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.0 %

Specification Test Results

About this item

 :
Boiling Pt:114°C
Flash Pt:22°C
UN:1993
ADR:3,II
 :102.1317 g/mol
EINECS:214-323-1
 :GHS02
 :GHS07
CAS Number:1120-97-4
Formula:C₅H₁₀O₂
MDL Number:MFCD00006569