

2-(2-Bromoethyl)-1,3-dioxane ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:33884-43-4
:
Formula:C₆H₁₁BrO₂
MDL Number:MFCD00006567
:195.06 g/mol
Boiling Pt:70°C
Flash Pt:97°C
EINECS:251-716-7
:GHS07
:GHS08
Specification Test Results
- Appearance:Colorless to Amber clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together