
2,4-Dichloro-5-sulphamoylbenzoic acid ≥97.0% (by HPLC, titration analysis)
TCIAD2541-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Neutralization titration):min. 97.0 %
- Solubility in Methanol:very faint turbidity
:
:270.09 g/mol
:220-358-3
:GHS05
:2736-23-4
:C₇H₅Cl₂NO₄S
:MFCD00007931