Order Entry
Italy
ContactUsLinkComponent
 
Lithium acetate 1 mol/l in aqueous solution
Catalog # R039
Supplier:  G-Biosciences
CAS Number:  
Lithium acetate 1 mol/l in aqueous solution
Catalog # R039
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :65.99 g/mol
Storage Temperature:Ambient
EINECS:208-914-3
CAS Number:546-89-4
Formula:H₃CCOOLi
MDL Number:MFCD00013057