



Methylamine 40% ∼9.8 mol/L in methanol
Supplier: TCI
Certificates
About this item
CAS Number:74-89-5
:
Formula:CH₅N
MDL Number:MFCD00008104
:31.06 g/mol
Storage Temperature:Ambient
EINECS:200-820-0
UN:3286
ADR:3,II
:GHS02
:GHS05
:GHS06
:GHS08
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Concentration (Neutralization titration):40.0 to 43.0 w/w%
Specification Test Results
Frequently Bought Together