


4,4'-Methylenedi-o-toluidine ≥97.0% (by titrimetric analysis)
TCIAD2046-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 94.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:152.0 to 159.0 deg-C
:
:226.3200 g/mol
:212-658-8
:3077
:9,III
:GHS07
:GHS08
:GHS09
:838-88-0
:C₁₅H₁₈N₂
:MFCD00126963