Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,4'-Methylenedi-o-toluidine
4,4'-Methylenedi-o-toluidine
4,4'-Methylenedi-o-toluidine
EHERC12194800
:
EHRENSTORFER
:
4,4'-Methylenedi-o-toluidine
EHERC12194800
:
EHRENSTORFER
:
Pack type:
Glass bottle for solids
Pk:
0,1 g
Seq:
3020
:
:
226.3200 g/mol
:
212-658-8
:
838-88-0
:
C₁₅H₁₈N₂
:
MFCD00126963