

3-(2,4-Dichlorophenoxy)propionic acid ≥98.0% (by titrimetric analysis)
Catalog # TCIAD1942-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Purity (Neutralization titration):min. 98.0 %
- Melting point:113.0 to 116.0 deg-C
Specification Test Results
About this item
:
:235.07 g/mol
Merck Index:14,03078
EINECS:204-390-5
UN:3077
ADR:9,III
:GHS05
:GHS07
CAS Number:120-36-5
Formula:C₉H₈Cl₂O₃
MDL Number:MFCD00002645