Order Entry
Italy
ContactUsLinkComponent
2-Methylbutyryl chloride 98%
Corrosive
Flammable
2-Methylbutyryl chloride 98%
Certificates
undefined
Corrosive
Flammable
2-Methylbutyryl chloride 98%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4160 to 1.4180 (20°C, 589 nm)
  • Titration RCOCl:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together