2-(Hydroxymethyl)-1,4-benzodioxane ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3663-82-9
:
Formula:C₉H₁₀O₃
MDL Number:MFCD00006822
Melting Pt:87…91 °C
:166.18 g/mol
Storage Temperature:Ambient
EINECS:222-921-9
Related Information
Frequently Bought Together