3-Methyl-1,5-pentanediol ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:4457-71-0
:
Formula:C₆H₁₄O₂
MDL Number:MFCD00039547
:118.18 g/mol
Boiling Pt:150°C
Flash Pt:146°C
EINECS:224-709-1
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together