
15-Hydroxypentadecanoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:4617-33-8
:
Formula:C₁₅H₃₀O₃
MDL Number:MFCD00039535
:258.40 g/mol
EINECS:225-026-1
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:86.0 to 89.0 deg-C
- Solubility in hot Methanol:almost transparency
Specification Test Results
Frequently Bought Together