Order Entry
Italy
ContactUsLinkComponent
1-(4-Chlorophenyl)cyclobutanecarboxylic acid 94%
Irritant
1-(4-Chlorophenyl)cyclobutanecarboxylic acid 94%
Catalog # ACRO170720250
CAS Number:  
undefined
Irritant
1-(4-Chlorophenyl)cyclobutanecarboxylic acid 94%
Catalog # ACRO170720250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Light yellow to purple Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    80 to 90 °C
  • HPLC:
    ≥92 %

Specification Test Results

About this item

 :
 :210.6568 g/mol
Melting Pt:80…90 °C
EINECS:256-847-3
 :GHS07
Storage Temperature:Ambient
CAS Number:50921-39-6
Formula:C₁₁H₁₁ClO₂
MDL Number:MFCD00001324